Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Miller's LB Agar, 1G Capsules, 5 KG
SDP

Catalog No. 501976939
Encompass_Preferred

Super convenient. Eliminate messy weighing. Consistant and accurate measurement. Convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments.

Simply add 40 capsules to one liter of water, sterilize by autoclaving for 15 minutes at 121°C. Capsules contain all necessary ingredients for maintenance and propagation of E. Coli.

Ingredients:

Casein Digest Peptone: 10g/L
Yeast Extract: 5g/L
Sodium Chloride: 10g/L
Agar:15g/L
TRIS/TRIS HCl: 1.5g/L

Catalog No. 50-197-6939 Supplier Research Products International Corp Supplier No. L240215000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Super convenient. Eliminate messy weighing. Consistant and accurate measurement. Convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments.

Simply add 40 capsules to one liter of water, sterilize by autoclaving for 15 minutes at 121°C. Capsules contain all necessary ingredients for maintenance and propagation of E. Coli.

Ingredients:

Casein Digest Peptone: 10g/L
Yeast Extract: 5g/L
Sodium Chloride: 10g/L
Agar:15g/L
TRIS/TRIS HCl: 1.5g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.