Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Luria Broth 1 Gram Buffered Capsules [Miller's LB Broth], 2.5 Kilograms
SDP

Catalog No. 50488755
Encompass_Preferred

High salt formula. Molecular Biology Grade. For maintenance and propagation of E. Coli.

RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.

Simply dissolve 25 capsules per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.0 ± 0.2 at 25°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.

Ingredients:

Casein digest of peptone: 10 g/L
Sodium Chloride: 10 g/L
Yeast Extract: 5 g/L
Tris/Tris HCl: 1.5 g/L

Catalog No. 50-488-755 Supplier Research Products International Corp Supplier No. L240412500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High salt formula. Molecular Biology Grade. For maintenance and propagation of E. Coli.

RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.

Simply dissolve 25 capsules per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.0 ± 0.2 at 25°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.

Ingredients:

Casein digest of peptone: 10 g/L
Sodium Chloride: 10 g/L
Yeast Extract: 5 g/L
Tris/Tris HCl: 1.5 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.