Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Luria Broth 1 Liter Buffered Capsules [Miller's LB Broth], 20 Capsules of 25 Grams, 500 Grams
SDP

Catalog No. 50488756
Encompass_Preferred

RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.

Simply dissolve 1 capsule per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.2 at 20°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.

For maintenance and propagation of E. Coli. High Salt Formula. Molecular Biology Grade.

Ingredients:

Tryptone: 10 g/L
Sodium Chloride: 10 g/L
Yeast Extract: 5 g/L
Tris/Tris HCl: 1.5 g/L

Catalog No. 50-488-756 Supplier Research Products International Corp Supplier No. L24045500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.

Simply dissolve 1 capsule per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.2 at 20°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.

For maintenance and propagation of E. Coli. High Salt Formula. Molecular Biology Grade.

Ingredients:

Tryptone: 10 g/L
Sodium Chloride: 10 g/L
Yeast Extract: 5 g/L
Tris/Tris HCl: 1.5 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.