Learn More
MSC Miller SkyGrip Temporary Horizontal Lifeline System, Galvanized Steel, 120 ft, 1 ea
Catalog No. 5000507166
Encompass
This temporary horizontal lifeline system is engineered for concrete applications, providing fall protection with a 120 ft. (36.6m) length. It is constructed from galvanized steel for corrosion resistance and durability. The system features a modular design for versatile use and includes a tension indicator to ensure accurate fall clearance requirements.
  • Maximum working load capacity of 310 lb.
  • Meets ANSI A10.32, CSA, and OSHA standards.
  • Includes (2) energy absorbers, (3) stanchions, cable, chain connector, hardware, intermediate pass-through bracket, line tension indicator, slider bracket, tie-back bracket, and turnbuckle.
  • Features a continuous pass-through design for 100% tie-off.
  • Stanchions serve as single-point anchorages.
  • Supports up to 5 workers on systems for concrete with a maximum of 2 workers per span.
Catalog No. 50-005-07166 Supplier MSC Supplier No. 77135473
This temporary horizontal lifeline system is engineered for concrete applications, providing fall protection with a 120 ft. (36.6m) length. It is constructed from galvanized steel for corrosion resistance and durability. The system features a modular design for versatile use and includes a tension indicator to ensure accurate fall clearance requirements.
  • Maximum working load capacity of 310 lb.
  • Meets ANSI A10.32, CSA, and OSHA standards.
  • Includes (2) energy absorbers, (3) stanchions, cable, chain connector, hardware, intermediate pass-through bracket, line tension indicator, slider bracket, tie-back bracket, and turnbuckle.
  • Features a continuous pass-through design for 100% tie-off.
  • Stanchions serve as single-point anchorages.
  • Supports up to 5 workers on systems for concrete with a maximum of 2 workers per span.