Learn More
MSC Honeywell Miller SkyGrip Temporary Horizontal Lifeline System, Galvanized Steel, 120 ft, 1 ea
Catalog No. 5000507396
Encompass
This temporary horizontal lifeline system is designed for fall arrest and travel restraint applications on steel structures. The 120 ft (36.6 m) system is engineered for quick installation and provides continuous horizontal mobility for users working at height in outdoor or harsh environments.
  • Includes three stanchions, two energy absorbers, hardware, a line tension indicator, turnbuckle, and intermediate stanchion pass-through bracket.
  • Features a galvanized steel lifeline compatible with beam flanges 4 in. to 18 in. (0.1 m to 0.5 m) wide and up to 2-1/4 in. (57.2 mm) thick.
  • Supports up to four workers per system with a maximum working load of 310 lb per person.
  • Corrosion-resistant design suitable for demanding work sites.
  • Meets OSHA, ANSI A10.32, and CSA safety standards when correctly installed.
Catalog No. 50-005-07396 Supplier MSC Supplier No. 77135481
This temporary horizontal lifeline system is designed for fall arrest and travel restraint applications on steel structures. The 120 ft (36.6 m) system is engineered for quick installation and provides continuous horizontal mobility for users working at height in outdoor or harsh environments.
  • Includes three stanchions, two energy absorbers, hardware, a line tension indicator, turnbuckle, and intermediate stanchion pass-through bracket.
  • Features a galvanized steel lifeline compatible with beam flanges 4 in. to 18 in. (0.1 m to 0.5 m) wide and up to 2-1/4 in. (57.2 mm) thick.
  • Supports up to four workers per system with a maximum working load of 310 lb per person.
  • Corrosion-resistant design suitable for demanding work sites.
  • Meets OSHA, ANSI A10.32, and CSA safety standards when correctly installed.