Learn More
Grainger MILLER Spectrum 375 X-TREME Plasma Cutr
Catalog No. 502749370
Encompass_Preferred
The Miller Electric Spectrum 375 Plasma Cutter is a handheld plasma cutting system that can cut metal up to 5/8 inch thick. It has a recommended output range of 15-40 amps and can cut at speeds up to 12 inches per minute. The unit weighs 37 lbs and has a dual flow piercing control and post flow cooling. It comes with a 12 foot torch lead and requires 240V 1-Phase input power. The Spectrum 375 uses compressed air and has a recommended pressure of 65-75 PSI.
Catalog No. 50-274-9370 Supplier Grainger Supplier No. 13Z845
The Miller Electric Spectrum 375 Plasma Cutter is a handheld plasma cutting system that can cut metal up to 5/8 inch thick. It has a recommended output range of 15-40 amps and can cut at speeds up to 12 inches per minute. The unit weighs 37 lbs and has a dual flow piercing control and post flow cooling. It comes with a 12 foot torch lead and requires 240V 1-Phase input power. The Spectrum 375 uses compressed air and has a recommended pressure of 65-75 PSI.