Learn More
Honeywell™ Miller™ Swivel Brackets for MightyLite™ Self-Retracting Lifelines
Catalog No. 19034879
Change view
Click to view available options
For Use With (Equipment):
Self retracting lifeline, 20 ft. and 30 ft. MightyLite™ unit, edge fall protection system
Self retracting lifeline, 50 ft. and 65 ft. MightyLite™ unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-034-879 Self retracting lifeline, 50 ft. and 65 ft. MightyLite™ unit
19-120-784 Self retracting lifeline, 20 ft. and 30 ft. MightyLite™ unit, edge fall protection system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-879 Supplier Honeywell™ Supplier No. 9083/
Reduce webbing and cable wear, and drag resistance on the worker
Model 9082 bracket fits 20 and 30 ft. (6 and 9m) MightyLite lifelines (Cat. nos. 19-034-675, -679; 19-080-849; and 19-999-490)
TRUSTED_SUSTAINABILITY
Specifications
Material Zinc-plated Steel, Stainless Steel
For Use With (Equipment) Self retracting lifeline, 50 ft. and 65 ft. MightyLite™ unit
Product Type Swivel Bracket