Learn More
Grainger MILLER Wireless Hand Control Belt Clip
Catalog No. 502751742
Encompass_Preferred
The Miller Electric Wireless Hand Control Belt is a wireless remote control system that allows welders to adjust welding parameters from their waist. It has an operating range of 300 feet and features an LCD display with 4 digit amperage readout, 2 digit voltage readout and weld timer. The system includes a transmitter belt pack with membrane switches, an antenna and receiver module, and connects via cables to Millermatic or Syncrowave power sources. This wireless control system provides more mobility and convenience for welders.
Catalog No. 50-275-1742 Supplier Grainger Supplier No. 13Z843
The Miller Electric Wireless Hand Control Belt is a wireless remote control system that allows welders to adjust welding parameters from their waist. It has an operating range of 300 feet and features an LCD display with 4 digit amperage readout, 2 digit voltage readout and weld timer. The system includes a transmitter belt pack with membrane switches, an antenna and receiver module, and connects via cables to Millermatic or Syncrowave power sources. This wireless control system provides more mobility and convenience for welders.