Learn More
Grainger MILLER XT30 Handheld Plasma Torch
Catalog No. 502749228
Encompass_Preferred
The Miller Electric Plasma Cutting Tip For Miller 814NC5 is a replacement tip for Miller plasma cutters. It is designed for use with the Miller Spectrum 875, 875 Auto-Line, and 625 X-TREME plasma cutting systems. This is a standard tip with a .030" orifice size. It provides good cut quality on mild steel up to 5/8" thickness. The tip is made from high quality copper for durability and improved heat transfer. Proper tip maintenance and replacement helps optimize cut quality and consumable life. This is an OEM part from Miller Electric.
Catalog No. 50-274-9228 Supplier Grainger Supplier No. 13Z846
The Miller Electric Plasma Cutting Tip For Miller 814NC5 is a replacement tip for Miller plasma cutters. It is designed for use with the Miller Spectrum 875, 875 Auto-Line, and 625 X-TREME plasma cutting systems. This is a standard tip with a .030" orifice size. It provides good cut quality on mild steel up to 5/8" thickness. The tip is made from high quality copper for durability and improved heat transfer. Proper tip maintenance and replacement helps optimize cut quality and consumable life. This is an OEM part from Miller Electric.