Learn More
Grainger MILLER XT40 Handheld Plasma Torch
Catalog No. 502749208
Encompass_Preferred
The Miller Electric XT40 Plasma Torch is a handheld plasma cutting torch designed for metal fabrication and repair. It has a recommended cut capacity of 5/8 inch mild steel and can cut stainless steel and aluminum. The XT40 uses compressed air to cut and requires an air compressor that provides 5-7 CFM at 90 PSI. It has an output of 40 amps and an input voltage of 240V. The torch comes with a 12 foot lead and features a spring loaded safety trigger for improved control. It is lightweight at 5 lbs and has a 2 year warranty. The XT40 plasma torch delivers clean, fast cuts in metal up to 5/8 inches thick.
Catalog No. 50-274-9208 Supplier Grainger Supplier No. 13Z851
The Miller Electric XT40 Plasma Torch is a handheld plasma cutting torch designed for metal fabrication and repair. It has a recommended cut capacity of 5/8 inch mild steel and can cut stainless steel and aluminum. The XT40 uses compressed air to cut and requires an air compressor that provides 5-7 CFM at 90 PSI. It has an output of 40 amps and an input voltage of 240V. The torch comes with a 12 foot lead and features a spring loaded safety trigger for improved control. It is lightweight at 5 lbs and has a 2 year warranty. The XT40 plasma torch delivers clean, fast cuts in metal up to 5/8 inches thick.