Learn More
Grainger MILLER XT40 Handheld Plasma Torch
Catalog No. 502749383
Encompass_Preferred
The Miller Electric XT40 Plasma Torch is a handheld plasma cutting torch designed for metal fabrication and repair. It has a recommended cut capacity of 5/8 inch mild steel and can cut stainless steel and aluminum. The XT40 uses compressed air to cut metal with a superheated plasma arc up to 20,000°F. It requires an air compressor and plasma cutter power supply to operate. The torch comes with a 12 foot lead and features an ergonomic handle with trigger start. It has replaceable electrode and nozzle consumable parts. The XT40 provides high cutting speed and quality cuts in a portable handheld plasma torch.
Catalog No. 50-274-9383 Supplier Grainger Supplier No. 13Z852
The Miller Electric XT40 Plasma Torch is a handheld plasma cutting torch designed for metal fabrication and repair. It has a recommended cut capacity of 5/8 inch mild steel and can cut stainless steel and aluminum. The XT40 uses compressed air to cut metal with a superheated plasma arc up to 20,000°F. It requires an air compressor and plasma cutter power supply to operate. The torch comes with a 12 foot lead and features an ergonomic handle with trigger start. It has replaceable electrode and nozzle consumable parts. The XT40 provides high cutting speed and quality cuts in a portable handheld plasma torch.