Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Millers LB Agar Hgh Salt 25KG
SDP

Catalog No. 502230464
Encompass_Preferred

For propagation and maintenance of E. Coli. High Salt Formula. Molecular Biology Grade.

  • Dissolve 40 grams in 1 liter distilled water
  • Adjust pH to 7.2
  • Sterilize by autoclaving for 15 minutes

Ingredients:
Peptone: 10g/l
Yeast: 5 g/l
Sodium Chloride: 10g/l
Agar: 15g/l

Catalog No. 50-223-0464 Supplier Research Products International Corp Supplier No. L2402025000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For propagation and maintenance of E. Coli. High Salt Formula. Molecular Biology Grade.

  • Dissolve 40 grams in 1 liter distilled water
  • Adjust pH to 7.2
  • Sterilize by autoclaving for 15 minutes

Ingredients:
Peptone: 10g/l
Yeast: 5 g/l
Sodium Chloride: 10g/l
Agar: 15g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.