Learn More
Research Products International Corp Luria Broth Granulated [Miller's LB Broth], 2 Kilogram
SDP
Catalog No. 501368335
Encompass_Preferred
For propagation and maintenance of E. Coli. High Salt Formula. Molecular Biology Grade.
Dissolve 25 grams in 1 liter distilled water and adjust the pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes. Material specially granulated to reduce dust and increase solubility rate.
Ingredients:
Tryptone:10 g/l
Sodium Chloride: 10 g/l
Yeast Extract: 5 g/l
Catalog No. 50-136-8335 Supplier Research Products International Corp Supplier No. L244002000.0
For propagation and maintenance of E. Coli. High Salt Formula. Molecular Biology Grade.
Dissolve 25 grams in 1 liter distilled water and adjust the pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes. Material specially granulated to reduce dust and increase solubility rate.
Ingredients:
Tryptone:10 g/l
Sodium Chloride: 10 g/l
Yeast Extract: 5 g/l