Learn More
Research Products International Corp Luria Broth, High Salt Formula, Powder [Miller's LB Broth], 1 Kilogram
SDP
Catalog No. 50213404
Encompass_Preferred
For propagation and maintenance of E. Coli. High Salt Grade. Molecular Biology Grade.
Dissolve 25 grams in 1 liter distilled water
Adjust the pH to 7.2
Sterilize by autoclaving for 15 minutes
Ingredients:
Casein digest of peptone: 10 g/l
Yeast: 5 g/l
Sodium Chloride: 10 g/l
Catalog No. 50-213-404 Supplier Research Products International Corp Supplier No. L240401000.0
For propagation and maintenance of E. Coli. High Salt Grade. Molecular Biology Grade.
Dissolve 25 grams in 1 liter distilled water
Adjust the pH to 7.2
Sterilize by autoclaving for 15 minutes
Ingredients:
Casein digest of peptone: 10 g/l
Yeast: 5 g/l
Sodium Chloride: 10 g/l