Learn More
Research Products International Corp Luria Broth, High Salt Formula, Powder [Miller's LB Broth], 500 Grams
SDP
Catalog No. 50213405
Encompass_Preferred
For propagation and maintenance of E. Coli. High Salt Grade. Molecular Biology Grade.
Dissolve 25 grams in 1 liter distilled water
Adjust the pH to 7.2
Sterilize by autoclaving for 15 minutes
Ingredients:
Casein digest of peptone: 10 g/l
Yeast: 5 g/l
Sodium Chloride: 10 g/l
Catalog No. 50-213-405 Supplier Research Products International Corp Supplier No. L24040500.0
For propagation and maintenance of E. Coli. High Salt Grade. Molecular Biology Grade.
Dissolve 25 grams in 1 liter distilled water
Adjust the pH to 7.2
Sterilize by autoclaving for 15 minutes
Ingredients:
Casein digest of peptone: 10 g/l
Yeast: 5 g/l
Sodium Chloride: 10 g/l