Learn More
MilliporeSigma™ Milli-Q™ EQ 7000 Foot Pedal
Catalog No. ZEQ7FTPDL Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Foot Pedal
Catalog No. Type
ZEQ7FTPDL Foot Pedal
1 options
Catalog No. ZEQ7FTPDL Supplier MilliporeSigma™ Supplier No. ZEQ7FTPDL
Add to Cart
Add to Cart
A hands-free dispensing accessory for Milli-Q™ EQ 7000 Water Purification Systems
  • Connect to the Q-POD™ Dispenser to allow for start and stop dispensing of ultrapure (Type 1) water with a simple push of the foot
  • When combined with Volumetric Dispense mode, one press on the pedal starts the production of ultrapure water at the point of dispense
  • Dispensing automatically stops at the set volume
  • Ideal for scientists who need both of their free hands when accessing purified water
  • Supports lab sanitary protocols since users do not need to touch the system for water access
TRUSTED_SUSTAINABILITY
Specifications
Type Foot Pedal
For Use With (Application) Hands-Free Dispensing
For Use With (Equipment) Milli-Q™ EQ 7000 Water Purification Systems