Learn More
MilliporeSigma Milli-Q™ EQ 7008/16 Ultrapure Water Purification Kit
Catalog No. EQ70XXPKIT Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
EQ70XXPKIT Tap
1 options
Catalog No. EQ70XXPKIT Supplier MilliporeSigma Supplier No. EQ70XXPKIT
Add to Cart
Add to Cart
For Milli-Q™ EQ 7008/16 systems to produce RO (Type 3) and ultrapure (Type 1) water. This product replaces EQ70XXPKT1.
  • For Milli-Q™ EQ 7008/16 ultrapure water purification systems
  • Optimally protects RO membranes (IPAK Gard™), removes trace ions and organics (Milli-Q™ polishing cartridges), and protect stored water from airborne contaminants (Tank Vent Filter)
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
Purification Method Ultrapure
Resin Type Ion Exchange Resin
Type Ultrapure Water Purification Kit