Learn More
MilliporeSigma™ Milli-Q™ EQ Wall Mounting Kit
Catalog No. WMEQ0RKT Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Milli-Q™ EQ 7000 Q-POD Dispenser
Milli-Q™ EQ 7000 Touchscreen
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
WMEQ0RKT Milli-Q™ EQ 7000 Q-POD Dispenser
WMEQ0DKT Milli-Q™ EQ 7000 Touchscreen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. WMEQ0RKT Supplier MilliporeSigma™ Supplier No. WMEQ0RKT
Add to Cart
Add to Cart
Save precious laboratory space by wall-mounting the accessories for your EQ 7000 Water Purification System.
The Q-POD™ Dispenser or HMI Touchscreen for the Milli-Q™ EQ 7000 Water Purification System can be wall-mounted up to 3 m from the system, saving precious space.
  • Includes all necessary components for convenient installation of the Q-POD™ Dispenser or HMI Touchscreen for the Milli-Q™ EQ 7000 Water Purification System
  • Optimizes installation flexibility of the system, allowing setup to adapt to ever-changing laboratories
TRUSTED_SUSTAINABILITY
Specifications
Type Wall Mounting Kit
For Use With (Equipment) Milli-Q™ EQ 7000 Q-POD Dispenser