Learn More
MilliporeSigma Milli-Q™ Polishing Kit for Purified Water
Catalog No. IPAKKIT00 Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Purified Water
Catalog No. Feedwater Source
IPAKKIT00 Purified Water
1 options
Catalog No. IPAKKIT00 Supplier MilliporeSigma Supplier No. IPAKKIT00
You must be signed in and have a business account to purchase online.
For Milli-Q™ IQ/EQ 7000 and EQ 7008/16 ultrapure water systems fed with purified water. This product replaces IPAKKITA1.
  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with one drain cap per cartridge, to remove excess water and reduce the weight of exhausted cartridges at disposal.
  • 78% of the parts suppliers for the cartridges included in this kit are ISO 14001 environmental management system-certified.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Purified Water
Purification Method Ultrapure
Resin Type Ion Exchange Resin
Type Polishing Kit