Learn More
MilliporeSigma Milli-Q™ Pressure Regulator
Catalog No. ZFMQPR3BT0 Shop All MilliporeSigma Products
Change view
Click to view available options
Type: Pressure Regulator
Catalog No.
ZFMQPR3BT0 Type: Pressure Regulator
1 options
Catalog No. ZFMQPR3BT0 Supplier MilliporeSigma Supplier No. ZFMQPR3BT0
Add to Cart
Add to Cart
Limits tap feed water pressure to 3 bars to ensure optimal RO membrane performance
  • Pressure Regulator limits tap feed water pressure to 3 bars to ensure optimal RO membrane performance of Milli-Q™ systems
TRUSTED_SUSTAINABILITY
Specifications
Type Pressure Regulator