Learn More
MilliporeSigma™ Milli-Q™ Reference Ultrapure Water Purification System
Catalog No. Z00QSV0US Shop All MilliporeSigma Products
Change view
Click to view available options
Includes:
US shipping kit (power cord, documentation)
Catalog No. Includes
Z00QSV0US US shipping kit (power cord, documentation)
1 options
Catalog No. Z00QSV0US Supplier MilliporeSigma™ Supplier No. Z00QSV0US
Includes
U.S. shipping kit (power cord, documentation)
Make ultrapure water delivery convenient with the Milli-Q™ Reference Ultrapure Water Purification System.
This system delivers ultrapure (Type 1) water customized to fit a broad range of research areas. System makes ultrapure water available in any laboratory, regardless of space limitations or the stringency of regulatory norms.
  • Designed to fit the height and shape of all laboratory glassware
  • Easily prepares solutions with the low flow function to precisely adjust the meniscus in volumetric flasks
Model Z00QSVC01
Includes TOC indicator
TRUSTED_SUSTAINABILITY
Specifications
Conductivity <100ppb
Contaminant Rejection (Microbiological) Less than1cfu/mL
Feedwater Source Elix, Reverse Osmosis, Deionized, Distilled
Flow Rate 50 and 2000 mL/min.
Product Water Type Type 1
Resistivity 18.2
Total Organic Carbon <5
Type Water Purification System
Includes US shipping kit (power cord, documentation)
Weight (English) 41 lb. Packaged, 42 lb. with water
Weight (Metric) 19 kg Packaged, 19.5 kg with water
Voltage 110/230 V