Learn More
MilliporeSigma Milli-Q™ SQ 200 Purification System Kit
Catalog No. ZSQ200U0TK Shop All MilliporeSigma Products
Change view
Click to view available options
Type: Water Purification System
Catalog No.
ZSQ200U0TK Type: Water Purification System
1 options
Catalog No. ZSQ200U0TK Supplier MilliporeSigma Supplier No. ZSQ200U0TK
Add to Cart
Add to Cart
Ultrapure (Type 1) water dispensing module; Includes one 3.5 L Milli-Q™ SQ Switch Tank.
  • The Milli-Q™ SQ 200 water purification system delivers ultrapure (Type 1) water from the SQ Switch 3.5 L tank filled with pure reverse osmosis (RO) water.
  • It benefits from a flexible handset dispenser to deliver ultrapure water. This dispensing module must be associated with a Milli-Q™ SQ 2Series family pure RO production station to refill its mobile SQ Switch tank.
  • Kit requires 1 Power Cord, Cartridge, Final Filter, and Installation (not included).
  • Features and Benefits:
    • Reliable 18.2 MΩ·cm resistivity ultrapure (Type 1) water quality is indicated at a glance by a colored LED
    • TOC is reduced by an integrated photo-oxidation UV lamp
    • Fast (as little as 6 min.) refilling of the mobile SQ Switch tank with freshly produced pure RO water due to the high-flow RO cartridge (up to 42 L/h) of the pure water production station; RO water recovery loop saves 50% of the rejected water; automatic RO membrane rinsing maintains its high performance.
    • Compact setup and self-guided installation manual and videos let you plug the system wherever is convenient for your lab activity and move it later if needed
    • Individual Twist and Lock cartridge replacement simplifies maintenance and limits running cost
    • Compact size plus water and energy savings support lab sustainability efforts
TRUSTED_SUSTAINABILITY
Specifications
Type Water Purification System