Learn More
MilliporeSigma Milli-Q™ SQ 200P Purification System
Catalog No. ZSQ200UPT0 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.22 μm
Catalog No. Pore Size
ZSQ200UPT0 0.22 μm
1 options
Catalog No. ZSQ200UPT0 Supplier MilliporeSigma Supplier No. ZSQ200UPT0
Add to Cart
Add to Cart
Request a Quote
Ultrapure (Type 1) water dispensing module from a pressurized purified water loop
  • Milli-Q™ SQ 200 Module Only.
  • Produces ultrapure water plumbed from a pretreated purified water source.
  • Does not include required Power Cord, Purification Cartridge, Final Filter, or Installation.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Pretreated
Flow Rate 1.6 L/min. Distribution Flow Rate (Ultrapure water from Handset Dispenser), 1.6 L/min. product water instant delivery rate (Ultrapure water from Handset Dispenser)
Pore Size 0.22 μm
Purification Method Membrane Filter