Learn More
MilliporeSigma Milli-Q™ SQ 240C Purification System
Catalog No. ZSQ240UCT0 Shop All MilliporeSigma Products
Change view
Click to view available options
Purification Method: Membrane Filter
Catalog No.
ZSQ240UCT0 Purification Method: Membrane Filter
1 options
Catalog No. ZSQ240UCT0 Supplier MilliporeSigma Supplier No. ZSQ240UCT0
Add to Cart
Add to Cart
Request a Quote
Combined pure RO water production station and ultrapure (Type 1) water dispensing module with flexible dispensing
  • Milli-Q™ SQ 240C Module Only.
  • Produces ultrapure water from tap water via Milli-Q SR 240 RO Module, SQ Switch 3.5L Tank, and Milli-Q SQ 200 Ultrapure Module.
  • Manual dispenser with variable flow rate.
  • Does not include required Power Cord, Purification Cartridges, SQ Switch 3.5L Tank, Final Filter, or Installation.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
Flow Rate 1.6 L/min. Distribution Flow Rate (Ultrapure water from Handset Dispenser), 1.6 L/min. product water instant delivery rate (Ultrapure water from Handset Dispenser), 42 L/hr. Make-up Flow Rate (for pure RO water), 6 L/min. Distribution Flow Rate (Pure RO water from SQ Gravity valve), 6 L/min. product water instant delivery rate (Pure RO water from SQ Gravity valve)
Pore Size 0.22 μm
Purification Method Membrane Filter