Description
- Milli-Q™ SQ 240CV Module Only. Produces ultrapure water from tap water via Milli-Q SR 240 RO Module, SQ Switch 3.5L Tank, and Milli-Q SQ 200 Ultrapure Module.
- Includes user-programmable volumetric dispensing head.
- Does not include required Power Cord, Purification Cartridges, SQ Switch 3.5L Tank, Final Filter, or Installation.
Specifications
Specifications
| Feedwater Source | Tap |
| Flow Rate | 1.6 L/min. Distribution Flow Rate (Ultrapure water from Preset Volume Dispenser), 1.6 L/min. product water instant delivery rate (Ultrapure water from Preset Volume Dispenser), 6 L/min. Distribution Flow Rate (Pure RO water from SQ Gravity valve), 6 L/min. product water instant delivery rate (Pure RO water from SQ Gravity valve) |
| Pore Size | 0.22 μm |
| Purification Method | Membrane Filter |