Learn More
MilliporeSigma Milli-Q™ SQ 240XL Purification System Kit
Catalog No. ZSQ240RXTK Shop All MilliporeSigma Products
Change view
Click to view available options
Product Water Type:
Ultrapure (Type 1)
Catalog No. Product Water Type
ZSQ240RXTK Ultrapure (Type 1)
1 options
Catalog No. ZSQ240RXTK Supplier MilliporeSigma Supplier No. ZSQ240RXTK
Add to Cart
Add to Cart
Request a Quote
Ultrapure Water Dispensing Module, two SQ Switch 3.5 L Tanks, one SQ 50 L Tank, SQ Gravity Valve
  • The Milli-Q™ SQ 240XL Purification System Kit is composed of the Milli-Q™ SR 240XL and the Milli-Q™ SQ 200 water purification systems.
  • The Milli-Q™ SR 240XL water purification system delivers pure reverse osmosis (RO) water to one mobile SQ Switch 3.5 L tank and one SQ 50 L tank. The SQ 50 L tank benefits from a Gravity valve (optional) to directly dispense pure RO water plus a high-flow valve to directly feed lab equipment with pure RO water.
  • The Milli-Q™ SQ 200 water purification system delivers ultrapure (Type 1) water from the SQ Switch 3.5 L tank filled with pure RO water. It benefits from a flexible handset dispenser to deliver ultrapure water.
  • Kit requires 2x Power Cords, Cartridges, Final Filter, and Installation (not included).
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2 x 3.5 L Tanks, 1 x 50 L Tank
Feedwater Source Potable Tap Water
Flow Rate ≤1.6 L/min. Distribution Flow Rate (Ultrapure water from Handset Dispenser), ≤12 L/min. Distribution Flow Rate (Pure RO water from high flow SQ 50 L tank valve), ≤42 L/hr. Make-up Flow Rate (Pure RO water), ≤6 L/min. Distribution Flow Rate (Pure RO water from SQ Gravity valve)
Product Water Type Ultrapure (Type 1)
TOC Monitor with UV Intensity Monitoring ≤5 ppb
Type Water Purification System