Description
- Milli-Q™ SR 240XL Module Only.
- Includes filling station for 1x 3L SQ Switch Tank ability to fill SQ XL 50L Tank.
- Produces reverse osmosis water when used with SQPAK RO and compatible Tanks.
- Does not include required Power Cord, Purification Cartridge, Tanks, or Installation.
Specifications
Specifications
| Type | Water Purification System |