Learn More
MilliporeSigma Millicell™ ERS 3.0 Digital Voltohmmeter
Catalog No. 09804177 Shop All MilliporeSigma Products
Change view
Click to view available options
Voltage:
0/1 V
Catalog No. Voltage
09-804-177 0/1 V
1 options
Catalog No. 09-804-177 Supplier MilliporeSigma Supplier No. MERS03000
Add to Cart
Add to Cart
Request a Quote
The Millicell Electrical Resistance System reliably measures transmembrane potential and resistance of endothelial/epithelial cells in culture.
  • Reliably measures membrane potential and resistance of epithelial and endothelial cells in culture
  • Qualitatively measures cell monolayer health and cell confluence
  • An Electroless Nickle Immersion Gold (ENIG) plate on each external facing electrode tip measures voltage
  • Small electrode size enables easy measurement of transepithelial voltage and the resistance of cells grown on microporous membranes
  • Comes with the Voltohmmeter, standard adjustable electrode, a Wi-Fi USB dongle, a power cord, a verification device and free 6-month trial subscription to Millicell™ Cloud account.
  • Each instrument is battery compatible but can run off only AC wall power in the absence of a battery.
TRUSTED_SUSTAINABILITY
Specifications
Type Digital Voltohmmeter
Voltage 0/1 V