Learn More
MilliporeSigma Millicell™ ERS 3.0 Verification Device
Catalog No. 09804169 Shop All MilliporeSigma Products
Change view
Click to view available options
Voltage:
0/1 V
Catalog No. Voltage
09-804-169 0/1 V
1 options
Catalog No. 09-804-169 Supplier MilliporeSigma Supplier No. MERS03VER
Add to Cart
Add to Cart
Verification probe for Millicell ERS 3.0 Voltohmmeter.
  • Reliably measures membrane potential and resistance of epithelial and endothelial cells in culture
  • Qualitatively measures cell monolayer health and cell confluence
  • Designed to test the Millicell™ ERS 3.0 Digital Voltohmmeter
TRUSTED_SUSTAINABILITY
Specifications
Type Verification Device
Voltage 0/1 V