Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Millipore Millicell™ Microwell Plates

Catalog No. MC96U4005 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
400 μm
600 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
MC96U4005 400 μm
MC96U6005 600 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MC96U4005 Supplier Millipore Supplier No. MC96U4005

A read-to-use platform for high-throughput and reproduciblity of organoid cultures.

Millicell Microwell Plates are a read-to-use platform for high-throughput and reproduciblity of organoid cultures. Based on an array of ultra-dense U-bottom shaped microwells (uW) in hydrogel, single organoids are robustly generated in each microcvity and grown in suspension-like culture without a solid extracellular matrix. With mulitple configurations available, there is a microwell size that will fit your organoid model.

Features and benefits:

  • O-ring with meniscus breaking features: For homogeneous seeding
  • Independent port: For medium change without disturbing the organoids
  • Multiple microwell sizes: Accommodate any organoid type
  • Biocompatible and cell repellent: For efficient aggregation of stem cells
  • Water-based: Allowing diffraction-less imaging
  • Tunable chemistry: For customization according to organoid needs
  • No solid matrix needed
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Pore Size 400 μm
Color Clear
Product Type Microwell Plates
Material Polystyrene
Volume (Metric) Well 200 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.