Learn More
MilliporeSigma Millicell™ Standard Electrode Probe
Catalog No. 09804171 Shop All MilliporeSigma Products
Change view
Click to view available options
Voltage:
0/1 V
Catalog No. Voltage
09-804-171 0/1 V
1 options
Catalog No. 09-804-171 Supplier MilliporeSigma Supplier No. MERS03SAP
Add to Cart
Add to Cart
Standard adjustable electrode for Millicell ERS 3.0 Digital Voltohmmeter which can be used with 6-, 12-, and 24-well inserts.
  • Fits into 3 different sizes of membrane inserts and receiver plates (6-well, 12-well, and 24-well) on the Millicell™ ERS 3.0 Digital VoltOhmMeter
  • Adjustable so that the depth of the electrode reaches the bottom of the well plate while the adjustable ring rests on top of the well
  • An Electroless Nickle Immersion Gold (ENIG) plate on each external facing electrode tip measures voltage.
  • Small electrode size enables easy measurement of transepithelial voltage and the resistance of cells grown on microporous membranes
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode
Voltage 0/1 V