Learn More
Millipore Millicell™ Ultra-low Attachment Plates
Catalog No. MC96ULA20 Shop All MilliporeSigma Products
Change view
Click to view available options
Well Shape:
Round Bottom (U-well)
Catalog No. Well Shape
MC96ULA20 Round Bottom (U-well)
1 options
Catalog No. MC96ULA20 Supplier Millipore Supplier No. MC96ULA20
Add to Cart
Add to Cart
Ultra-low Attachment (ULA) Plates provide quality three-dimensional cell culture, to enable spheroid culturing of specific cell types.
Millicell™ Ultra-low Attachment (ULA) Plates provide you with superior quality three-dimensional cell culture, to enable spheroid culturing of your specific cell type. ULA plates that promote scaffold-free, selfassembly of spheroid formation. The plates are precoated with a unique, ultra-hydrophilic polymer which enables spontaneous spheroid formation of uniform size and shape. The 96-well U bottom ULA plates have high optical clarity, making them highly suitable for bright field imaging and confocal microscopy.
Features and benefits:
  • Non-binding surface for cells to facilitate natural spheroid formation
  • Uniform single spheroid/EB formation in each well
  • Spheroid assay formation and analysis in the same plate
  • High optical clarity plates for imaging
  • Stable, non-cytotoxic and cell non-adhesion surface
  • Easy handling, compatible with liquid robotic system
  • Suitable for brightfield imaging and fluorescence microscopy
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Color Clear
Product Type Ultra-low Attachment Plates
Material Polystyrene
Volume (Metric) Well 300 μL , 250 μL Recommended
No. of Wells 96
Sterility Sterile
Surface Treatment Hydrophilic
Well Shape Round Bottom (U-well)
Sterilization Method Gamma-irradiated