Learn More
MilliporeSigma Milliflex funnel, 100ml, 24 per pack
Supplier:  MilliporeSigma MXGSWG124
Encompass
Milliflex filter funnel unit, 0.22micrometer, MCE, white, gridded surface, 100mL funnel, sterile, 24/PK.
Catalog No. NC9412438