Learn More
MilliporeSigma MILLIFLEX AGAR CASSETTE 48/PK
Catalog No. NC9510167
Encompass
Pre-filled Milliflex Cassettes, product name: Centrimide, target microorganisms, Pseudomonas aeruginosa, for use with the Milliflex system, 48/PK.
Catalog No. NC9510167 Supplier MilliporeSigma Supplier No. MXSMCET48
Pre-filled Milliflex Cassettes, product name: Centrimide, target microorganisms, Pseudomonas aeruginosa, for use with the Milliflex system, 48/PK.