Learn More
MilliporeSigma Milliflex Cassette prefilled with Tryptic Soy Agar with Polysorbate 80 and Lecithin, 48 tests
Catalog No. NC0159471
Encompass
Milliflex Cassette prefilled with Tryptic Soy Agar is used in Bioburden testing and general microbiological analysis. It is commonly used in determining the efficiency of the sanitation of containers, equipment and surfaces as well as for water miscible cosmetic products. It is widely used in pharmaceutical, cosmetic and brewing industry.
Catalog No. NC0159471 Supplier MilliporeSigma Supplier No. MXSMTLP48
Milliflex Cassette prefilled with Tryptic Soy Agar is used in Bioburden testing and general microbiological analysis. It is commonly used in determining the efficiency of the sanitation of containers, equipment and surfaces as well as for water miscible cosmetic products. It is widely used in pharmaceutical, cosmetic and brewing industry.