Learn More
MilliporeSigma MILLIFLEX FILT.FUNNEL 24/PK
Supplier:  MilliporeSigma MXHABG124
Encompass
Milliflex filter funnel unit, 100mL funnel, sterile, black, gridded, pore size 0.45micrometer, MCE, 24/PK.
Catalog No. NC9385166