Learn More
MilliporeSigma MILLIFLEX OASIS TRYPTIC SOY
Catalog No. NC1835545
Encompass
Milliflex Oasis Tryptic Soy media plates for detection of aerobic and anaerobic bacteria in water
Catalog No. NC1835545 Supplier MilliporeSigma Supplier No. MMSMCTS48
Milliflex Oasis Tryptic Soy media plates for detection of aerobic and anaerobic bacteria in water