Learn More
MilliporeSigma Milliflex Rapid Reagent Kit, 100 tests
Catalog No. NC9849107
Encompass
An integral component in the Milliflex Rapid Microbiology Detection and Enumeration system, the Milliflex Rapid AutoSpray Station applies an equal volume of two reagents across a membrane surface in order to detect living microorganisms.
Catalog No. NC9849107 Supplier MilliporeSigma Supplier No. MXRPBLRST
An integral component in the Milliflex Rapid Microbiology Detection and Enumeration system, the Milliflex Rapid AutoSpray Station applies an equal volume of two reagents across a membrane surface in order to detect living microorganisms.