Learn More
Cole-Parmer Extech 380560 High Resolution Milliohm Meter; 110 VAC
Supplier:  Cole-Parmer 2002415
Encompass
The benchtop meter provides high resolution down to 0.01 mΩ using 4-wire Kelvin clip test leads.It features seven ranges for wide 20mO to 20kO low resistance measurements, a large 0.8" digits 1999 count display, automatic zero, and 5V test voltage.
Catalog No. 50-992-972