Learn More
INNOVATIVE BIOCHIPS Million Microwell Device 5PK
Catalog No. nc1637882
Encompass_Preferred
The Million Microwell Device (MMD) is a high-throughput and low-cost platform for single-cell isolation and RNA sequencing. The MMD is made of biocompatible PDMS material and contains 1,000,000 high-quality cylindrical microwells of 30 µm in diameter and 33 µm in depth. The optimized size of microwells is compatible with efficient single-cell isolation of most cells. The MMD allows to easily generating up to 100,000 pairings of single cells and single barcoded magnetic beads into single microwells. The single cell-bead pairings can be clearly identified through bight field, phase contrast, and fluorescence imaging under a x10, x20, and x40 objective. In microwells, the isolated single cells are lysed and their mRNA are captured by the paired barcoded mRNA capture beads for subsequent transcriptome sequencing.
Catalog No. NC1637882 Supplier INNOVATIVE BIOCHIPS Supplier No. H4MMD5PK
The Million Microwell Device (MMD) is a high-throughput and low-cost platform for single-cell isolation and RNA sequencing. The MMD is made of biocompatible PDMS material and contains 1,000,000 high-quality cylindrical microwells of 30 µm in diameter and 33 µm in depth. The optimized size of microwells is compatible with efficient single-cell isolation of most cells. The MMD allows to easily generating up to 100,000 pairings of single cells and single barcoded magnetic beads into single microwells. The single cell-bead pairings can be clearly identified through bight field, phase contrast, and fluorescence imaging under a x10, x20, and x40 objective. In microwells, the isolated single cells are lysed and their mRNA are captured by the paired barcoded mRNA capture beads for subsequent transcriptome sequencing.