Learn More
EMD MILLIPORE 3P BIOPHARM MILLIPAK 100 UNSTERILIZED 3/PK
Supplier:  EMD MILLIPORE 3P BIOPHARM MPGL10CA3
Encompass
Millipak-100 filter units, 0.22um, 14mm (9/16 in.) hose barb inlet and outlet, nonsterile, typical applications include: sterile filtration of parenterals, diagnostics and buffers, media and biologicals
Catalog No. NC9330381