Learn More
EMD MILLIPORE 3P BIOPHARM MILLIPAK 200 0.22UM STR.FLT3PK
Catalog No. NC9438573
Encompass
Millipak-200 , 0.22micrometer, 14 mm ( 9/16 in. ) hose barb inlet and outlet, nonsterile, 3/PK.
Catalog No. NC9438573 Supplier EMD MILLIPORE 3P BIOPHARM Supplier No. MPGL20CA3
Millipak-200 , 0.22micrometer, 14 mm ( 9/16 in. ) hose barb inlet and outlet, nonsterile, 3/PK.