Learn More
MilliporeSigma Mossel Broth acc. EP+USP, 10/pk
Catalog No. NC1744714
Encompass
Mossel Broth is suitable for microbiological examination of non-sterile pharmaceutical products, foodstuff and other materials. It is designed for selective enrichment of bile-tolerant, gram-negative bacteria and particularly Enterobacteriaceae.
This item is not returnable. View return policy
Catalog No. NC1744714 Supplier MilliporeSigma Supplier No. 1464630010
Mossel Broth is suitable for microbiological examination of non-sterile pharmaceutical products, foodstuff and other materials. It is designed for selective enrichment of bile-tolerant, gram-negative bacteria and particularly Enterobacteriaceae.