Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MilliSentials™ Lab Adaptable Labels

Catalog No. 09735144 Shop All MilliporeSigma Products
Change view
Click to view available options
Dimensions (L x W):
1.125 x 1.72 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
09-735-144 1.125 x 1.72 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-735-144 Supplier MilliporeSigma™ Supplier No. MISELADLA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MilliSentials™ Lab Adaptable labels have been custom designed and optimized for use throughout the laboratory, withstanding temperatures from -196°C (liquid nitrogen) to 100°C (boiling water).

  • Labels provide resistance to harsh chemical solvents (including ethanol, DMSO, xylene)
  • Labels have been designed with perforations, allowing one size to fit across a wide range of laboratory vessels including microcentrifuge tubes, cryovials, petri dishes, cell culture plates, and reagent bottles
  • Single label size can adapt across multiple laboratory vessels
  • Temperature and chemical solvent resistant
TRUSTED_SUSTAINABILITY

Specifications

Adhesive Type Hot-melt, Self-adhesive
Color White
Display Blank
Finish Matte
Format Roll
Printable Sides Single-side Printable
Product Type Label
Sterility Non-sterile
Display Text None
For Use With (Application) Laboratory Sample Labeling
For Use With (Equipment) MilliSentials Lab Labeling System
Includes Roll of 1615 Labels
Resolution 300 dpi
Shape Rectangle with Square Corners
Size (English) 1 in. Core (Media Roll Size)
Dimensions (L x W) 1.125 x 1.72 in.
Length (English) 1.125 in.
Width (English) 1.72 in.
Width (Metric) 43.6 mm
Show More Show Less

Unless otherwise stated, products are intended for research use only and are not to be used for any other purpose, including but not limited to unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption or application to humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.