Learn More
Description
The Millipore MilliSolve Filtration System has been designed for the filtration of liquid chromatography buffers and solvents under vacuum. The system uses a 0.45 μm membrane filter to eliminate particles which can shorten column life. Vacuum filtration with the Millipore MilliSolve system also removes a large portion of dissolved gasses from buffers, reducing the risk of air bubbles interfering with LC instruments. With automatic and continuous filtration, there is no need to add liquid into a funnel as filtration progresses. Filtration occurs in a closed system, which is important when filtering hazardous fluids. If open funnel filtration is required, the 300 mL funnel replaces the vacuum cover.
This item is a kit composed of the following items:
XX1614705 - Millipore Ground Joint Flask, 2L, Conical bottom
XX1514702 - Millipore Glass Base and Cap, 47 mm, Glass frit support
XX1614701 - Millipore Glass cover, 47 mm, For use with Millipore MilliSolve Kit
XX1014703 - Millipore Spring Clamp, 47 mm, Aluminum
XT1200000 - Millipore Tubing for Solvent/Buffer, PTFE, 70 cm
XX1014704 - Millipore Glass funnel, 300 mL, 47 mm, Borosilicate, Ground glass seal
ALIGNED05 - Millipore Alignment Guide, 47 mm, 5/PK
Features & Benefits:
Automatic, continuous solvent and buffer filtration eliminates the need to pour during filtration
- 2L receiving flask includes plug and tubing for storage, eliminating filtered buffer transfer, reducing labor and lessening contamination risk
- Conical 2L flask bottom allows access to all filtered solvent or buffer
Applications: Mobile Phase Preparation, Buffer Filtration
Specifications
Specifications
| Diameter (Metric) Inner | 12.5 cm |
| Includes | 2 L Flask, 47 mm dia. filter, Borosilicate Glass, Anodized Aluminum clamp, 120 in. PTFE tubing, Stainless Steel forceps, Glass vacuum support base with tube cap |
| Product Type | Laboratory Filter Holders or Cyclones |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.