Learn More
EMD MILLIPORE 3P BIOPHARM Millistak+ HC Pro Pod Depth Filter | C0SP media series | 0.014 m2 surface area | 1/4 in. Hose Barb connection
Supplier:  EMD MILLIPORE 3P BIOPHARM MC0SP014H1
Encompass
This is used in primary and secondary clarification (direct harvest; centrate).
Catalog No. NC1761532