Learn More
EMD MILLIPORE 3P BIOPHARM MILLISTAK+ POD FILTER GRADE X0
Supplier:  EMD MILLIPORE 3P BIOPHARM MX0HC027H1
Encompass
Millistak+ Pod Depth Filter, X0HC media series, 0.027m2 surface area, 1/4 in. Hose Barb connection, Secondary clarification of bioreactor harvests, primarily for cell cultures
Catalog No. NC0084768