Learn More
EMD MILLIPORE 3P BIOPHARM MILLISTAK+REGISTERED POD X0HC
Supplier:  EMD MILLIPORE 3P BIOPHARM MX0HC01FS1
Encompass
Millistak+ HC Pod Depth Filter, X0HC media series, 0.11m2 surface area, Secondary clarification of bioreactor harvests, primarily for cell cultures
Catalog No. NC0087280