Learn More
Covaris Inc MILLITUBE 1ML AFA FIBER (100)
SDP
Catalog No. NC0597431
Encompass
12x12mm tube with AFA Fiber ; PTFE seal cap (volume 1ml).; Rated for 1MHz, 150W PIP Case quantity available milliTUBE 1 ml AFA Fiber Case (1000) 520131
Catalog No. NC0597431 Supplier Covaris Inc Supplier No. 520130
12x12mm tube with AFA Fiber ; PTFE seal cap (volume 1ml).; Rated for 1MHz, 150W PIP Case quantity available milliTUBE 1 ml AFA Fiber Case (1000) 520131