Learn More
Covaris Inc MILLITUBE 1ML WITH AFA FIBER
SDP
Catalog No. NC0586946
Encompass
12x12 mm tube with AFA Fiber & PTFE seal cap (Volume 1 ml). Rated for 1 MHz, 150W PIP. 24/Pk.
Catalog No. NC0586946 Supplier Covaris Inc Supplier No. 520135
12x12 mm tube with AFA Fiber & PTFE seal cap (Volume 1 ml). Rated for 1 MHz, 150W PIP. 24/Pk.